GC Analysis of Phthalate Esters on Equity®-5
GC Analysis of Phthalate Esters on Equity®-5
Conditions
column Equity-5, 30 m x 0.25 mm I.D., 0.25 µm (28089-U)
oven 40 °C (1 min), 10 °C/min to 325 °C
carrier gas helium, 1.3 mL/min constant flow
sample 50 ng on-column of a custom phthalate ester mix
injection 1 µL, splitless (0.75 min)
liner 4 mm I.D., single taper
inj. temp. 250 °C
detector FID
Description
Categories Analytical Chromatography, GC Applications, Esters, Phthalate Esters
Featured Industry Chemicals and Industrial Polymers
Cosmetics, Personal Care, and Cleaning Products
Environmental
Food and Beverages
Legal Information Equity is a registered trademark of Sigma-Aldrich Co. LLC
Other Notes Complementary products:
Syringes
Vials
Gas Purification and Gas Management
Inlet Liners, Glass Wool, and Specialized Hand Tools
Septa and Specialized Hand Tools
Column Ferrules, Nuts, and Specialized Hand Tools
suitability application for GC
Analyte
  • Benzyl benzoate
  • Bis(2-butoxyethyl) phthalate
  • Bis(2-ethoxethyl) phthalate
  • Bis(2-ethylhexyl) phthalate
  • Bis(2-methoxyethyl) phthalate
  • Bis(4-methyl-2-pentyl) phthalate
  • Butylbenzylphthalate
  • Diamyl phthalate
  • Dibutyl phthalate
  • Dicyclohexyl phthalate
  • Diethyl phthalate
  • Diisobutylphthalate
  • Dimethylphthalate
  • Di-n-Hexyl phthalate
  • Di-n-Nonyl phthalate
  • Di-n-Octyl phthalate
  • Hexyl-2-ethylhexyl phthalate