GC Analysis of Phthalate Esters in Corn Oil (Spiked) on SLB®-5ms after SPME with a 100 μm PDMS Fiber
GC Analysis of Phthalate Esters in Corn Oil (Spiked) on SLB®-5ms after SPME with a 100 μm PDMS Fiber
Conditions
sample/matrix corn oil spiked with phthalate esters at 500 µg/kg
desorption process 260 °Cfor 4 min
extraction headspace, 95 °C (measured in headspace)
SPME fiber 100 µm PDMS (57300-U)
column F.S. CAP. SLB-5ms 20 m x 0.18 mm I.D., 0.18 µm (28564-U)
oven 60 °C (1 min), 10 °C/min to 330 °C (10 min)
carrier gas helium, 0.6 mL/min, constant
sample Corn oil
liner 0.75 mm I.D. SPME
inj. temp. 260 °C
Description
Analysis Note In this application, SPME was used for the analysis of phthalate esters from oily samples, specifically the oil-based flavor packets included with ramen noodle kits. Spiked samples of corn oil and flavored oils included with ramen noodle kits were analyzed using high temperature SPME headspace extraction using a 100 μm PDMS fiber which was then directly desorbed into the GC/MS system. Calibration standards were prepared in corn oil, and spiked samples of chicken and beef oil-based flavors were analyzed. The SLB-5ms GC columns provide consistently low-bleed, inert, efficient, and durable separations.
Categories Analytical Chromatography, SPME Applications, GC Applications, Agriculture, Volatiles, Phthalate Esters
Featured Industry Agriculture
Food and Beverages
Legal Information SLB is a registered trademark of Sigma-Aldrich Co. LLC
Other Notes Complementary products:
Syringes
Vials
Gas Purification and Gas Management
Inlet Liners, Glass Wool, and Specialized Hand Tools
Septa and Specialized Hand Tools
Column Ferrules, Nuts, and Specialized Hand Tools
suitability application for SPME, application for GC