GC Analysis of Polynuclear Aromatic Hydrocarbons (PAHs) in Grilled Hamburger on SPB®-608 (20 m x 0.18 mm I.D., 0.18 μm) after QuEChERS Cleanup using Supel™ QuE Z-Sep, Fast GC Analysis
GC Analysis of Polynuclear Aromatic Hydrocarbons (PAHs) in Grilled Hamburger on SPB®-608 (20 m x 0.18 mm I.D., 0.18 μm) after QuEChERS Cleanup using Supel™ QuE Z-Sep, Fast GC Analysis
Conditions
sample preparation Dispersive (QuEChERS)
sample/matrix grilled hamburger spiked with PAHs at 100 ng/g
extraction tube Sodium chloride/magnesium sulfate (55294-U)
extraction process add 25 mL acetonitrile, contents of unbuffered salt tube (55294-U); shake for 5 minutes; centrifuge for 5 minutes at 3200 rpm
clean-up process transfer 3 mL of acetonitrile layer into a Supel QuE Z-Sep cleanup tube (55403-U); shake 1 minute; centrifuge for 3 minutes at 3400 rpm; remove 1 mL supernatant for GC-MS analysis
clean-up tube Supel QuE Z-Sep tube (55403-U)
column SPB-608, 20 m x 0.18 mm I.D. x 0.18 µm
oven 60 °C (1 min), 25 °C/min to 275 °C, 10 °C/min to 300 °C (13 min)
carrier gas helium, 1.5 mL/min constant flow
sample grilled hamburger extract
injection 1 µLsplitless (1 min.)
liner 4 mm I.D. FocusLiner™ with taper
inj. temp. 265 °C
detector MS-SIM
MSD interface 300 °C
Description
Analysis Note Sample pretreatment: Place 5 g ground hamburger sample into 50 mL centrifuge tube (55248-U). Spike with 100 ng/g target PAHs and internal standards. Let sit 10 min. Add 5 mL water and homogenize., This application demonstrates the use of dispersive SPE or QuEChERS for the extraction and cleanup of grilled hamburger for GC-MS analysis of polynuclear aromatic hydrocarbons (PAHs). Z-Sep sorbent was used in the cleanup step and found to reduce background and yield recoveries >80% for a majority of the PAHs tested.
Categories Analytical Chromatography, SPE Applications, GC Applications, Hydrocarbons, PAHs
Featured Industry Food and Beverages
Legal Information SPB is a registered trademark of Sigma-Aldrich Co. LLC, Supel is a trademark of Sigma-Aldrich Co. LLC, FocusLiner is a trademark of SGE Analytical Science Pty Ltd.
Other Notes Complementary products:
Syringes
Vials
Gas Purification and Gas Management
Inlet Liners, Glass Wool, and Specialized Hand Tools
Septa and Specialized Hand Tools
Column Ferrules, Nuts, and Specialized Hand Tools
suitability application for GC, application for SPE