GC Analysis of 'Positive List' Chemicals (Mix 5) on SLB®-5ms
GC Analysis of 'Positive List' Chemicals (Mix 5) on SLB®-5ms
Conditions
column SLB-5ms, 30 m x 0.25 mm I.D., 0.25 µm (28471-U)
oven 80 °C (2 min), 5 °C/min to 300 °C (4 min)
carrier gas helium, 1 mL/min constant
sample each pesticide at 20 ppm in acetone
injection 1.0 µLsplitless (1 min)
inj. temp. 250 °C
MSD interface 300 °C
scan range m/z 40-450
Description
Categories Analytical Chromatography, GC Applications, Herbicides, Pesticides
Featured Industry Agriculture
Food and Beverages
Legal Information SLB is a registered trademark of Sigma-Aldrich Co. LLC
Other Notes Complementary products:
Syringes
Vials
Gas Purification and Gas Management
Inlet Liners, Glass Wool, and Specialized Hand Tools
Septa and Specialized Hand Tools
Column Ferrules, Nuts, and Specialized Hand Tools
suitability application for GC
Analyte
  • Ametryn
  • Bifenazate
  • Biphenyl
  • Bitertanol
  • Bromacil
  • Bromuconazole
  • Captan
  • Captofol
  • Chinomethionate
  • Chlorpropylate
  • Chlorthalonil
  • Clofentezine
  • CNP
  • Dichlofluanid
  • Dichlofluanid metabolite
  • Diclocymet
  • Famoxadone
  • Fenbuconazole
  • Fenoxaprop-ethyl
  • Fludioxonil
  • Folpet
  • Furametpyr
  • Furametpyr metabolite
  • Hexaconazole
  • Iprodione
  • Iprodione metabolite
  • Mefenacet
  • Methoprene
  • Methyl 3,4-dichloro carbanilate (Swep)
  • Metribuzin
  • Nitralin
  • Nitrofen
  • Oxabetrinil
  • Oxadixyl
  • Pentoxazone
  • Phenothiol
  • Piperonyl butoxide
  • Propanil
  • Propargite
  • Pyrazoxyfen
  • Pyriproxyfen
  • Pyroquilon
  • Silafluofen
  • Simetryn
  • Tetramethrin
  • Thiocyclam
  • Tolylfluanid
  • Tolylfluanid metabolite
  • Trichlamide
  • α-BHC
  • β-BHC
  • γ-BHC
  • δ-BHC