GC Analysis of QA Test Mix on SLB®-5ms, Three Different Phase Lots
GC Analysis of QA Test Mix on SLB®-5ms, Three Different Phase Lots
Conditions
column SLB-5ms, 30 m x 0.25 mm I.D., 0.25 µm (28471-U)
oven 110 °C (14 min), 15 °C/min to 325 °C (15 min)
carrier gas helium, 30 cm/sec @ 110 °C
sample Nonpolar Column Test Mix, each analyte at 500 µg/mL in methylene chloride
injection 1.0 µL100:1 split
liner 4 mm I.D. split, cup design
inj. temp. 250 °C
detector FID, 360 °C
Description
Categories Analytical Chromatography, GC Applications, Hydrocarbons
Featured Industry General Analytical
Legal Information SLB is a registered trademark of Sigma-Aldrich Co. LLC
Other Notes Complementary products:
Syringes