GC Analysis of QA Test Mix on SLB®-IL59 using Air as Carrier Gas, Comparison to a Polyethylene Glycol (PEG) Column
GC Analysis of QA Test Mix on SLB®-IL59 using Air as Carrier Gas, Comparison to a Polyethylene Glycol (PEG) Column
Conditions
column SLB-IL59, 30 m x 0.25 mm I.D., 0.20 µm (28891-U)
column PEG phase, 30 m x 0.25 mm I.D., 0.25 µm
oven 50 °C (2 min), 4 °C/min to 200 °C (15 min)
carrier gas compressed air, 16 psi constant
sample Programmed Test Mix
injection 1 µL50:1 split
inj. temp. 250 °C
detector 250 °C
Description
Categories Analytical Chromatography, GC Applications, Hydrocarbons
Featured Industry General Analytical