GC Analysis of Residual Solvents on Equity®-5
GC Analysis of Residual Solvents on Equity®-5
Conditions
column Equity-5, 30 m x 0.53 mm I.D., 5.0 µm (28279-U)
oven 40 °C (6 min), 2 °C/min to 100 °C
carrier gas helium, 20 cm/sec @ 40 °C
sample 5 ng on-column of a 24 component solvent standard in DMSO
injection 1 µL, 10:1 split
liner split, cup design
inj. temp. 225 °C
detector FID, 250 °C
Description
Categories Analytical Chromatography, GC Applications, Solvents
Featured Industry Pharmaceutical (small molecule)
Legal Information Equity is a registered trademark of Sigma-Aldrich Co. LLC
Other Notes Complementary products:
Syringes
Vials
Gas Purification and Gas Management
Inlet Liners, Glass Wool, and Specialized Hand Tools
Septa and Specialized Hand Tools
Column Ferrules, Nuts, and Specialized Hand Tools
suitability application for GC
Analyte
  • 1,1,1-Trichloroethane
  • 1,1-Dichlorothylene
  • 1,2-Dichloroethane
  • 1,4-Dioxane
  • 2-Butanone
  • 2-Propanol
  • 4-Methyl-2-pentanone
  • Acetone
  • Acetonitrile
  • Benzene
  • Carbon tetrachloride
  • Chloroform
  • Dimethyl formamide
  • Ethanol
  • Ethyl acetate
  • Ethyl ether
  • Freon 113
  • Hexane
  • Methanol
  • Methyl tert-butyl ether
  • Methylene chloride
  • Tetrahydrofuran (THF)
  • Toluene
  • Trichloroethylene