GC Analysis of Spearmint Essential Oil on SUPELCOWAX® 10
GC Analysis of Spearmint Essential Oil on SUPELCOWAX® 10
Conditions
column SUPELCOWAX 10, 30 m x 0.25 mm I.D., 0.25 µm (24079)
oven 75 °C (4 min), 4 °C/min to 200 °C (5 min)
carrier gas helium, 25 cm/sec @ 155 °C
injection 0.2 µL, 100:1 split
detector FID
Description
Categories Analytical Chromatography, GC Applications, Essential Oils
Featured Industry Flavors and Fragrances
Legal Information SUPELCOWAX is a registered trademark of Sigma-Aldrich Co. LLC
Other Notes Complementary products:
Syringes
Vials
Gas Purification and Gas Management
Inlet Liners, Glass Wool, and Specialized Hand Tools
Septa and Specialized Hand Tools
Column Ferrules, Nuts, and Specialized Hand Tools
suitability application for GC
Analyte
  • 1,8-Cineole
  • 3-Octanol
  • 3-Octyl acetate
  • Carvone
  • cis-Carveol
  • cis-Carvyl acetate
  • cis-Jasmone
  • Dihydrocarvone
  • Germacrene D
  • Linalool
  • L-Limonene
  • L-Menthone
  • Myrcene
  • p-Cymene
  • Sabinene
  • Terpinen-4-ol
  • Terpinolene
  • trans-Carveol
  • trans-Dihydrocarvyl acetate
  • trans-Farnesene
  • trans-Sabinene hydrate
  • Viridiflorol
  • α-Pinene
  • α-Terpinene
  • α-Terpineol
  • β-Bourbonene
  • β-Caryophyllene
  • β-Pinene
  • γ-Terpinene