GC Analysis of Sterols in Soybean Oil on SAC™-5
GC Analysis of Sterols in Soybean Oil on SAC™-5
Conditions
column SAC-5, 30 m x 0.25 mm I.D., 0.25 µm (24156)
oven 265 °C
carrier gas helium, 20 cm/sec @ 265 °C
sample soybean oil
injection 1 µL, 100:1 split
inj. temp. 300 °C
detector FID, 300 °C
Description
Categories Analytical Chromatography, GC Applications, Sterols
Featured Industry Agriculture
Food and Beverages
Legal Information SAC is a trademark of Sigma-Aldrich Co. LLC
Other Notes Complementary products:
Syringes
Vials
Gas Purification and Gas Management
Inlet Liners, Glass Wool, and Specialized Hand Tools
Septa and Specialized Hand Tools
Column Ferrules, Nuts, and Specialized Hand Tools
suitability application for GC
Analyte
  • 5-α-Cholestane
  • Campesterol
  • Stigmasterol
  • β-Sitosterol