GC Analysis of Sweet Orange Essential Oil on SLB®-5ms (10 m x 0.10 mm I.D., 0.10 μm), Fast GC Analysis
GC Analysis of Sweet Orange Essential Oil on SLB®-5ms (10 m x 0.10 mm I.D., 0.10 μm), Fast GC Analysis
Conditions
column SLB-5ms, 10 m x 0.10 mm I.D., 0.10 µm (28465-U)
oven 40 °C, 50 °C/min to 320 °C
carrier gas hydrogen, 81.5 cm/sec constant
sample sweet orange essential oil in hexane
injection 0.4 µL, 300:1 split
inj. temp. 320 °C
detector FID, 320 °C
Description
Categories Analytical Chromatography, GC Applications, Essential Oils
Featured Industry Flavors and Fragrances
Legal Information SLB is a registered trademark of Sigma-Aldrich Co. LLC
Other Notes Complementary products:
Syringes
Vials
Gas Purification and Gas Management
Inlet Liners, Glass Wool, and Specialized Hand Tools
Septa and Specialized Hand Tools
Column Ferrules, Nuts, and Specialized Hand Tools
suitability application for GC
Analyte
  • (-)-Perilla aldehyde (perillaldehyde)
  • (-)-trans-Caryophyllene
  • (-)-α-Copaene
  • (-)-β-Pinene
  • (+)-Sabinene
  • (+)-Terpinen-4-ol
  • (+)-Valencene
  • (+)-δ-Cadinene
  • ®-(+)-Citronellal
  • ®-(+)-Limonene
  • 1-Octanol
  • 3,7-Dimethyl-2,6-octadienyl acetate
  • 3-Carene
  • Bicyclogermacrene
  • Camphene
  • cis-Limonene oxide
  • Decanal
  • Geranial
  • Geranyl acetate
  • Germacrene D
  • Lauric aldehyde
  • Linalool
  • Myrcene
  • Neral
  • Neryl acetate
  • Nonanal
  • Octanal
  • p-Cymene
  • Perilla alcohol
  • Terpinolene
  • trans-2-Decenal
  • trans-Limonene oxide
  • trans-β-Farnesene
  • trans-β-Ocimene
  • Undecanal
  • α-Huµlene
  • α-Phellandrene
  • α-Pinene
  • α-Terpinene
  • α-Terpineol
  • α-Thujene
  • β-Copaene
  • γ-Terpinene