GC Analysis of Tall Oil Fatty Acid Methyl Esters on SP™-2380
GC Analysis of Tall Oil Fatty Acid Methyl Esters on SP™-2380
Conditions
column SP-2380, 30 m x 0.25 mm I.D., 0.20 µm (24110-U)
oven 170 °C, 4 °C/min to 260 °C (4 min)
carrier gas helium, 20 cm/sec @ 185C
sample Tall oil methyl esters in methylene chloride
injection 2 µL, split 100:1
inj. temp. 250 °C
detector FID, 280 °C
Description
Categories Analytical Chromatography, GC Applications, Fatty Acid Methyl Esters (FAMEs)
Featured Industry Chemicals and Industrial Polymers
Legal Information SP is a trademark of Sigma-Aldrich Co. LLC
Other Notes Complementary products:
Syringes