GC Analysis of trans-1,2-Cyclohexanediol Enantiomers on Astec® CHIRALDEX® B-PH
GC Analysis of trans-1,2-Cyclohexanediol Enantiomers on Astec® CHIRALDEX® B-PH
Conditions
column Astec CHIRALDEX B-PH, 30 m x 0.25 mm I.D., 0.12 µm (71023AST)
oven 120 °C
carrier gas helium, 30 psi
sample peaks 1 & 2: trans-1,2-cyclohexanediol enantiomers
inj. temp. 250 °C
detector FID, 250 °C
Description
Categories Analytical Chromatography, GC Applications, Diols, Enantiomers, Glycols
Featured Industry Chemicals and Industrial Polymers
Legal Information Astec is a registered trademark of Sigma-Aldrich Co. LLC, CHIRALDEX is a registered trademark of Sigma-Aldrich Co. LLC
Other Notes Complementary products:
Syringes