GC Analysis of trans C18:1 FAMEs in Partially Hydrogenated Vegetable Oil (PHVO) on SLB®-IL111 after HPLC Fractionation using Silver Ion
GC Analysis of trans C18:1 FAMEs in Partially Hydrogenated Vegetable Oil (PHVO) on SLB®-IL111 after HPLC Fractionation using Silver Ion
Conditions
column SLB-IL111, 100 m x 0.25 mm I.D., 0.20 µm (29647-U)
oven 168 °Cisothermal
carrier gas hydrogen, 1 mL/min
sample trans fraction of partially hydrogenated vegetable oil FAMEs, prepared using Ag-ion fractionation
injection 1 µL, 100:1 split
liner 4 mm I.D., split liner with cup (2051001)
inj. temp. 250 °C
detector FID, 250 °C
Description
Categories Analytical Chromatography, GC Applications, Fatty Acid Methyl Esters (FAMEs)
Featured Industry Agriculture
Food and Beverages
Legal Information SLB is a registered trademark of Sigma-Aldrich Co. LLC
Other Notes Complementary products:
Syringes