GC Analysis of Volatile Flavor Compound Enantiomers on Astec® CHIRALDEX® G-TA
GC Analysis of Volatile Flavor Compound Enantiomers on Astec® CHIRALDEX® G-TA
Conditions
column Astec CHIRALDEX G-TA, 30 m x 0.25 mm I.D., 0.12 µm (73033AST)
oven 60 °C, 2 °C/ min to 160 °C (hold 20 min)
carrier gas helium, 30 cm/sec, set at 75 °C
injection 1 µL, split 100:1
inj. temp. 250 °C
detector FID, 250 °C
Description
Categories Analytical Chromatography, GC Applications, Flavors, Fragrances, Enantiomers, Volatiles
Featured Industry Flavors and Fragrances
Legal Information Astec is a registered trademark of Sigma-Aldrich Co. LLC, CHIRALDEX is a registered trademark of Sigma-Aldrich Co. LLC
suitability application for GC
Analyte
  • Carvone
  • Methyl lactate
  • Methyl mandelate
  • α-Ionone
  • α-Phenylethanol
  • β-Butyrolactone
  • δ-Octanolactone