GC Analysis of Wieland-Miescher Ketone Enantiomers on Astec® CHIRALDEX® G-TA
GC Analysis of Wieland-Miescher Ketone Enantiomers on Astec® CHIRALDEX® G-TA
Conditions
column Astec CHIRALDEX G-TA, 30 m x 0.25 mm I.D., 0.12 µm (73033AST)
carrier gas helium, 35 psi
gradient 110 °C, 5 °C/min to 160 °C
sample peak 1: S(+)-Wieland-Miescher Ketonepeak 2: R(-)-Wieland-Miescher Ketone
inj. temp. 250 °C
detector FID, 250 °C
Description
Analysis Note Elution order reversal on CHIRALDEX G-BP.
Categories Analytical Chromatography, GC Applications, Ketones, Enantiomers
Featured Industry Chemicals and Industrial Polymers
Legal Information Astec is a registered trademark of Sigma-Aldrich Co. LLC, CHIRALDEX is a registered trademark of Sigma-Aldrich Co. LLC
Other Notes Complementary products:
Syringes