GCxGC Analysis of a 59-Component Industrial Solvent Mix on SLB®-5ms x SLB-IL60, Non-Polar to Polar Strategy
GCxGC Analysis of a 59-Component Industrial Solvent Mix on SLB®-5ms x SLB-IL60, Non-Polar to Polar Strategy
Conditions
column (1D) SLB-5ms, 30 m x 0.25 mm I.D., 0.25 µm (28471-U)
column (2D) SLB-IL60, 1 m x 0.10 mm I.D., 0.08 µm
oven (modulator) temp. offset 30 °C, modulation period 3 sec
oven (1D) 60 °C, 10 °C/min to 225 °C (5 min)
oven (2D) 75 °C, 10 °C/min to 240 °C (2 min)
carrier gas helium, 1.2 mL/min
sample 59-Component industrial solvent mix
injection 1 µL100:1 split
inj. temp. 250 °C
detector TOFMS, mass range 35-450 m/z, acquisition rate 200 spectra/sec, ion source temp. 230 °C
Description
Analysis Note LECO Pegasus 4D GCxGC/TOFMS
Categories Analytical Chromatography, GC Applications, Solvents