GCxGC Analysis of B20 Biodiesel on SLB®-5ms x SLB-IL60, Non-Polar to Polar Strategy
GCxGC Analysis of B20 Biodiesel on SLB®-5ms x SLB-IL60, Non-Polar to Polar Strategy
Conditions
column (1D) SLB-5ms, 30 m x 0.25 mm I.D., 0.25 µm (28471-U)
column (2D) SLB-IL60, 1 m x 0.10 mm I.D., 0.08 µm
oven (1D) 40 °C (2 min), 4 °C/min to 270 °C (20 min)
oven (2D) 55 C (2 min), 4 °C/min to 280 °C (20 min)
oven (modulator) temp. offset 30 °C; modulation period 3 sec
carrier gas helium, 1.0 mL/min
sample B20 biodiesel
injection 1 µL50:1 split
inj. temp. 250 °C
detector TOFMS, mass range 35-450 m/z, acquisition rate 200 spectra/sec, ion source temp. 250 °C
Description
Categories Analytical Chromatography, GC Applications, PAHs, Hydrocarbons, Fatty Acid Methyl Esters (FAMEs)
Featured Industry Biofuels