US EPA Method 8082: GC Analysis of Aroclor 1016/1260 (PCBs) on Equity®-5
US EPA Method 8082: GC Analysis of Aroclor 1016/1260 (PCBs) on Equity®-5
Conditions
column Equity-5, 30 m x 0.25 mm I.D., 0.25 µm (28089-U)
oven 100 °C (2 min), 15 °C/min to 160 °C, 5 °C/min to 300 °C (10 min)
carrier gas helium, 30 cm/sec @ 100 C
sample Aroclor Mix 1 standard at 75 ppb with surrogates at 7.5 ppb (46846-U)
injection 2 µL, splitless (0.5 min)
liner Splitless double taper, unpacked
inj. temp. 225 °C
detector ECD, 310 °C
Description
Categories Analytical Chromatography, GC Applications, Dioxins/PCBs/PBDEs
Featured Industry Environmental
Legal Information Equity is a registered trademark of Sigma-Aldrich Co. LLC
Other Notes Complementary products:
Syringes
Vials
Gas Purification and Gas Management
Inlet Liners, Glass Wool, and Specialized Hand Tools
Septa and Specialized Hand Tools
Column Ferrules, Nuts, and Specialized Hand Tools
suitability application for GC
Analyte
  • 2,4,5,6-Tetrachloro-m-xylene
  • Aroclor 1016
  • Aroclor 1260
  • Decachlorobiphenyl