US EPA Method TO-10/IP-8 and ASTM® D4861: GC Analysis of Pesticides on SPB®-608 after Collection/Desorption using ORBO™-1000
US EPA Method TO-10/IP-8 and ASTM® D4861: GC Analysis of Pesticides on SPB®-608 after Collection/Desorption using ORBO™-1000
Conditions
sample/matrix polyurethane foam (PUF) spiked with 250 ng each pesticide
adsorbent tube ORBO-1000 (20557)
column SPB-608, 30 m x 0.53 mm I.D., 0.5 µm film (25312)
oven 150 °C (1 min) to 270 °C at 4 °C/min
carrier gas helium, 7 mL/min
injection 1 µL
detector ECD
Description
Categories Analytical Chromatography, GC Applications, Air Monitoring Applications, Pesticides
Featured Industry Environmental
Industrial Hygiene
General description This is an application, not a product for sale.
See "Related Products" to find associated products for sale.
For more information, email: Supelco Technical Service
Legal Information SPB is a registered trademark of Sigma-Aldrich Co. LLC, ASTM is a registered trademark of American Society for Testing and Materials, ORBO is a trademark of Sigma-Aldrich Co. LLC
Other Notes Complementary products:
Syringes
Vials
Gas Purification and Gas Management
Inlet Liners, Glass Wool, and Specialized Hand Tools
Septa and Specialized Hand Tools
Column Ferrules, Nuts, and Specialized Hand Tools
suitability application for air monitoring, application for GC
Analyte
  • 2,4,5-Trichlorophenol
  • 2,4-D methyl ester
  • 4,4′-DDE (1,1-Dichloro-2,2-bis(p-chlorophenyl)ethylene)
  • 4,4′-DDT
  • Aldrin
  • Captan
  • Chlorothalonil
  • Chlorpyrifos
  • Dichlorvos
  • Dieldrin
  • Heptachlor
  • Heptachlor epoxide
  • Hexachlorobenzene
  • Methoxychlor
  • Mirex
  • Octachloronaphthalene
  • Pentachlorobenzene
  • Pentachlorophenol
  • Ronnel
  • trans-Nonachlor
  • α-BHC
  • β-BHC
  • γ-BHC