US EPA Method TO-17: GC Analysis of Volatiles on VOCOL® after Collection/Desorption using Air Toxics Tube
US EPA Method TO-17: GC Analysis of Volatiles on VOCOL® after Collection/Desorption using Air Toxics Tube
Conditions
sample/matrix 10-50 ng each analyte introduced onto tube (calibration standard, Cat. No. 41900)
adsorbent tube Air Toxics (glass) (25086)
column VOCOL, 60 m x 0.25 mm I.D., 1.5 µm film (24154)
oven 37 °C (4 min) to 200 °C at 4 °C/min, hold 10 min
carrier gas helium, 2 mL/min
detector FID
Description
Categories Analytical Chromatography, GC Applications, Air Monitoring Applications, Volatiles
Featured Industry Environmental
Industrial Hygiene
General description This is an application, not a product for sale.
See "Related Products" to find associated products for sale.
Or, for more information, email: Supelco Technical Service
Legal Information VOCOL is a registered trademark of Sigma-Aldrich Co. LLC
Other Notes Complementary products:
Syringes
Vials
Gas Purification and Gas Management
Inlet Liners, Glass Wool, and Specialized Hand Tools
Septa and Specialized Hand Tools
Column Ferrules, Nuts, and Specialized Hand Tools
suitability application for air monitoring, application for GC
Analyte
  • 1,1,1-Trichloroethane
  • 1,1,2,2-Tetrachloroethane
  • 1,1,2-Trichloroethane
  • 1,1-Dichloroethane
  • 1,1-Dichloroethene
  • 1,2,4-Trichlorobenzene
  • 1,2,4-Trimethylbenzene
  • 1,2-Dibromoethane
  • 1,2-Dichlorobenzene
  • 1,2-Dichloroethane
  • 1,2-Dichloropropane
  • 1,3,5-Trimethylbenzene
  • 1,3-Dichlorobenzene
  • 1,4-Dichlorobenzene
  • Benzene
  • Bromomethane
  • Carbon tetrachloride
  • Chlorobenzene
  • Chloroform
  • Chloromethane
  • cis-1,2-Dichloroethene
  • cis-1,3-Dichloropropene
  • Ethyl chloride
  • Ethylbenzene
  • Halocarbon 113
  • Halocarbon 114
  • Halocarbon 114
  • Halocarbon 12
  • Hexachloro-1,3-butadiene
  • Methylene chloride
  • m-Xylene
  • o-Xylene
  • p-Xylene
  • Styrene
  • Tetrachloroethene
  • Toluene
  • trans-1,3-Dichloropropene
  • Trichloroethene
  • Vinyl chloride