HPLC Analysis of 2,3-Dihydroxy-3-Phenylpropionic Acid Enantiomers on CHIROBIOTIC R
HPLC Analysis of 2,3-Dihydroxy-3-Phenylpropionic Acid Enantiomers on CHIROBIOTIC R
Conditions
column CHIROBIOTIC R, 25 cm x 4.6 mm I.D., 5 µm particles (13024AST)
column temp. ambient
mobile phase [A] 0.1% ammonia, pH 4.1 with formic acid; [B] methanol; (50:50, A:B)
flow rate 1 mL/min
sample 2,3-dihydroxy-3-phenyl-propionic acid isomers:(A) racemate A (4.85 and 6.95 min)(B) racemate B (5.33 and 6.29 min)
injection 10 µL
detector UV, 258 nm
Description
Categories Analytical Chromatography, Chiral Applications, Chemicals and Industrial Polymers, Organic Acids
Featured Industry Chemicals and Industrial Polymers
Pharmaceutical (small molecule)