HPLC Analysis of 2(4-Chlorophenoxy) Propionic Acid Enantiomers on Astec® CHIROBIOTIC® T
HPLC Analysis of 2(4-Chlorophenoxy) Propionic Acid Enantiomers on Astec® CHIROBIOTIC® T
Conditions
column CHIROBIOTIC T, 25 cm x 4.6 mm I.D., 5 µm particles (12024AST)
column temp. 25 °C
mobile phase 0.1 wt% ammonium formate in methanol
flow rate 1 mL/min
sample 2 mg/mL in methanol
injection 5 µL
detector UV, 235 nm
Description
Categories Analytical Chromatography, Chiral Applications, Environmental, Pesticides & Herbicides, All Environmental Applications, Herbicides
Featured Industry Agriculture