HPLC Analysis of 3',4'-Methylenedioxy-α-pyrrolidinopropiophenone (MDPPP) Enantiomers on Astec® CHIROBIOTIC® V2
HPLC Analysis of 3',4'-Methylenedioxy-α-pyrrolidinopropiophenone (MDPPP) Enantiomers on Astec® CHIROBIOTIC® V2
Conditions
column Astec CHIROBIOTIC V2 15 cm x 4.6 mm I.D., 5 µm particles (15023AST)
column temp. 20 °C
mobile phase [A] methanol; [B] water; [C] acetic acid; [D] ammonium hydroxide; (95:5:0.1:0.02, A:B:C:D)
flow rate 1 mL/min
pressure 1100 psi (70 bar)
sample 100 µg/mL in methanol
injection 5 µL
detector UV, 251 nm
Description
Analysis Note 3’,4’-Methylenedioxy-a-pyrrolidinopropiophenone, also known as MDPPP, is a synthetic cathinone found in numerous bath salt blends under the street name NRG-1. This designer drug first emerged in Germany in the late 1990s and early 2000s as an ingredient in imitation ecstasy pills. Astec CHIROBIOTIC V2, based on vancomycin CSP, provided resolution of the enatiomers. CRMs from Cerillliant provided reliable identification and quantification.
Categories Analytical Chromatography, Drugs of Abuse
Featured Industry Forensics and Toxicology