HPLC Analysis of Acetic Acid on SeQuant ZIC-HILIC
HPLC Analysis of Acetic Acid on SeQuant ZIC-HILIC
Conditions
column SeQuant ZIC-HILIC, PEEK, 150 x 4.6 mm, 5 µm, 200 Ã… (1.50455.0001)
column temp. ambient
mobile phase [A] acetonitrile; [B] ammonium acetate; (80:20, A:B)
flow rate 1.0 mL/min
injection 50 µL
detector 210 nm
Description
Categories Analytical Chromatography, Organic Acids