HPLC Analysis of Aflatoxins on Ascentis® Express C18
HPLC Analysis of Aflatoxins on Ascentis® Express C18
Conditions
column Ascentis Express C18, 10 cm x 2.1 mm I.D., 2.7 µm particles (53823-U)
column temp. 35 °C
mobile phase [A] 5 mM ammonium formate, pH 2.5 (titrated with formic acid); [B] methanol
gradient 5 to 100% B in 10 min
flow rate 0.4 mL/min
pressure 4714 psi (325 bar)
sample 0.15 - 30 mg/L in 90:10, water:acetonitrile
injection 5 µL
detector APCI (+)
Description
Analysis Note This application demonstrates the suitability of Ascentis Express C18 for the efficient separation of various toxins.
Categories Analytical Chromatography, Mycotoxins