HPLC Analysis of Aminopterin and Related Compounds on Ascentis® RP-Amide
HPLC Analysis of Aminopterin and Related Compounds on Ascentis® RP-Amide
Conditions
column Ascentis RP-Amide, 15 cm x 4.6 mm I.D., 5 µm particles (565352-U)
column temp. 35 °C
mobile phase [A] 10 mM ammonium formate, pH 3.0; [B] acetonitrile; (90:10, A:B)
flow rate 1.0 mL/min
injection 10 µL
detector UV, 280 nm
Description
Analysis Note This application demonstrates the suitability of the Ascentis RP-Amide for the analysis of aminopterin, folic acid and methotrexate. Structures along with the optimized chromatogram obtained on the Ascentis RP-Amide are presented.
Categories Analytical Chromatography, Antineoplastics