HPLC Analysis of Amyloid β-Protein Fragment 10-35 on Discovery® BIO Wide Pore C18
HPLC Analysis of Amyloid β-Protein Fragment 10-35 on Discovery® BIO Wide Pore C18
Conditions
column Discovery BIO Wide Pore C18, 25 cm x 4.6 mm I.D., 5 µm particles (568223-U)
column temp. 30 °C
mobile phase [A] 0.1% TFA in water; [B] 0.1% TFA in water:0.1% TFA in acetonitrile; (25:75)
flow rate 1.3 mL/min
injection 10 µL, 1 mg/mL in water
detector UV, 215 nm
Description
Categories Analytical Chromatography, Proteins
Featured Industry Life Science and Biopharma
Legal Information Discovery is a registered trademark of Sigma-Aldrich Co. LLC