HPLC Analysis of Aniline Homologs on Discovery® C18
HPLC Analysis of Aniline Homologs on Discovery® C18
Conditions
column Discovery C18, 15 cm x 4.6 mm I.D., 5 µm particles (504955)
column temp. 30 °C
mobile phase [A] methanol; [B] water (60:40, A:B)
flow rate 1 mL/min
detector UV, 254 nm
Description
Categories Analytical Chromatography, Dyes, Analgesics
Featured Industry Chemicals and Industrial Polymers
Legal Information Discovery is a registered trademark of Sigma-Aldrich Co. LLC