HPLC Analysis of Animal Feed Additives on Ascentis® Phenyl
HPLC Analysis of Animal Feed Additives on Ascentis® Phenyl
Conditions
column Ascentis Phenyl, 15 cm x 4.6 mm I.D., 5 µm particles (581616-U)
column temp. 35 °C
mobile phase [A] 10 mM ammonium formate (pH 3.0 with formic acid); [B] methanol; (78:22, A:B)
flow rate 1.0 mL/min
sample as indicated in mobile phase A
injection 5 µL
detector UV, 225 nm
Description
Categories Analytical Chromatography, Preservatives
Featured Industry Agriculture