HPLC Analysis of Antihistamines on Discovery® C8
HPLC Analysis of Antihistamines on Discovery® C8
Conditions
column Discovery C8, 15 cm x 4.6 mm I.D., 5 µm particles (59353-U)
column temp. 25 °C
mobile phase [A] acetonitrile; [B] 25 mM potassium phosphate, pH 2.3; (8:92, A:B)
flow rate 2 mL/min
injection 2 µL
detector UV, 254 nm
Description
Categories Analytical Chromatography, Pharmaceutical Analysis, Antihistamine
Featured Industry Pharmaceutical (small molecule)
Legal Information Discovery is a registered trademark of Sigma-Aldrich Co. LLC