HPLC Analysis of Antihypertensive Drugs (β-Calcium Channel Blockers and ACE Inhibitor) on Discovery® RP-AmideC16
HPLC Analysis of Antihypertensive Drugs (β-Calcium Channel Blockers and ACE Inhibitor) on Discovery® RP-AmideC16
Conditions
column Discovery RP-AmideC16, 15 cm x 4.6 mm I.D., 5 µm particles (505013)
column temp. 35 °C
mobile phase [A] methanol; [B] 25 mM KH2PO4, pH 7.0 (40:60, A:B)
flow rate 1 mL/min
injection 5 µL
detector UV, 214 nm
Description
Categories Analytical Chromatography, Pharmaceutical Analysis, Antihypertensives
Featured Industry Pharmaceutical (small molecule)
Legal Information Discovery is a registered trademark of Sigma-Aldrich Co. LLC