HPLC Analysis of Antipsychotic Drugs on Ascentis® RP-Amide
HPLC Analysis of Antipsychotic Drugs on Ascentis® RP-Amide
Conditions
column Ascentis RP-Amide, 15 cm x 4.6 mm I.D., 5 µm particles (565324-U)
column temp. 35 °C
mobile phase [A] 10 mM monobasic potassium phosphate (pH 3.0 with phosphoric acid); [B] methanol; (40:60, A:B)
flow rate 1 mL/min
sample 50 µg/mL each in mobile phase
injection 10 µL
detector UV, 220 nm
Description
Categories Analytical Chromatography, Pharmaceutical Analysis, Antipsychotics