HPLC Analysis of Antiretroviral Drugs on Ascentis® RP-Amide
HPLC Analysis of Antiretroviral Drugs on Ascentis® RP-Amide
Conditions
column Ascentis RP-Amide, 15 cm x 4.6 mm I.D., 5 µm particles (565324-U)
column temp. 35 °C
mobile phase [A] 25 mM ammonium phosphate (pH 5.5); [B] acetonitrile
flow rate 1.0 mL/min
sample as indicated in 25 mM ammonium phosphate (pH 5.5)
injection 10 µL
detector UV, 220 nm
Description
Categories Analytical Chromatography, Antiretrovirals