HPLC Analysis of Antithrombin III from Human Plasma on TSKgel® Heparin-5PW
HPLC Analysis of Antithrombin III from Human Plasma on TSKgel® Heparin-5PW
Conditions
column TSKgel Heparin-5PW, 7.5 cm x 7.5 mm I.D., 10 µm particles (813064)
gradient 0.15 M NaCl in 0.02 M Tris-HCl, pH 7.5, step gradient to 0.5 M NaCl at 30 min, step gradient to 1.5 M NaCl at 60 min
flow rate 1 ml/min
sample human plasma, 500 µl
detector UV, 280 nm
Description
Categories Analytical Chromatography, Proteins
Featured Industry Clinical
Life Science and Biopharma
Legal Information TSKgel is a registered trademark of Tosoh Corporation
Other Notes Data provided by Tosoh Bioscience LLC.