HPLC Analysis of Anxiolytic Drugs on Ascentis® Phenyl
HPLC Analysis of Anxiolytic Drugs on Ascentis® Phenyl
Conditions
column Ascentis Phenyl, 15 cm x 4.6 mm I.D., 5 µm particles (581616-U)
column temp. 35 °C
mobile phase [A] 10 mM ammonium acetate (pH 5.5 with acetic acid); [B] acetonitrile; (40:60, A:B)
flow rate 1 mL/min
injection 1 µL
detector UV, 254 nm
Description
Categories Analytical Chromatography, Antidepressants