HPLC Analysis of Atrolactic Acid Enantiomers on Astec® CHIROBIOTIC® R
HPLC Analysis of Atrolactic Acid Enantiomers on Astec® CHIROBIOTIC® R
Conditions
column CHIROBIOTIC R, 25 cm x 4.6 mm I.D., 5 µm particles (13024AST)
column temp. 25 °C
mobile phase 0.1 wt% ammonium acetate in methanol
flow rate 1 mL/min
sample 5 mg/mL in methanol
injection 2 µL
detector UV, 254 nm
Description
Categories Analytical Chromatography, Chiral Applications, Organic Acids, Chemicals and Industrial Polymers, Food & Beverage, All Food & Beverage Applications