HPLC Analysis of Benzodiazepines on Ascentis® Express C18, Acetonitrile Mobile Phase
HPLC Analysis of Benzodiazepines on Ascentis® Express C18, Acetonitrile Mobile Phase
Conditions
column Ascentis Express C18, 10 cm x 3 mm I.D., 2.7 µm particles (53814-U)
column temp. 35 °C
mobile phase [A] water; [B] acetonitrile; (66:34, A:B)
flow rate 0.6 mL/min
sample 0.1 g/L each in 20% methanol
injection 2 µL
detector UV, 250 nm
Description
Categories Analytical Chromatography, Clinical, Pharmaceutical Analysis, Anticonvulsant/antiepiletic/antiseizure, Antipsychotics, antiepileptic/antiseizure, antipsychotics
Featured Industry Clinical
Pharmaceutical (small molecule)