HPLC Analysis of Benzoin Enantiomers on Astec® Cellulose DMP
HPLC Analysis of Benzoin Enantiomers on Astec® Cellulose DMP
Conditions
column Astec Cellulose DMP, 15 cm x 4.6 mm I.D., 5 µm particles (51098AST)
column temp. 25 °C
mobile phase [A] heptane; [B] IPA; [C] TFA; (90:10:0.1, A:B:C)
flow rate 0.5 mL/min
sample 2 mg/mL in mobile phase
injection 2 µL
detector UV, 254 nm
Description
Categories Analytical Chromatography, Ketones, Antibiotics, Antimicrobials
Featured Industry Chemicals and Industrial Polymers
Pharmaceutical (small molecule)
Legal Information Astec is a registered trademark of Sigma-Aldrich Co. LLC