HPLC Analysis of Bovine Liver Extract on Mono Q® 5/50 GL
HPLC Analysis of Bovine Liver Extract on Mono Q® 5/50 GL
Conditions
column Mono Q 5/50 GL, 5 cm x 5 mm I.D., 10 µm particles (5-4807)
mobile phase [A] 50 mM diethanolamine, pH 8.8; [B] 50 mM diethanolamine + 1.0M sodium chloride, pH 8.8
flow rate 1.0 ml/min
injection 100 muL1.6 mg protein
detector UV, 280 nm
Description
Categories Analytical Chromatography, Proteins
Featured Industry Life Science and Biopharma
Legal Information Mono Q is a registered trademark of GE Healthcare