HPLC Analysis of Bupivacaine Enantiomers on Astec® CHIROBIOTIC® V2, Acetate Mobile Phase
HPLC Analysis of Bupivacaine Enantiomers on Astec® CHIROBIOTIC® V2, Acetate Mobile Phase
Conditions
column CHIROBIOTIC V2, 25 cm x 4.6 mm I.D., 5 µm particles (15024AST)
column temp. 25 °C
mobile phase [A] methanol; [B] 20 mM ammonium acetate, pH 4.1; (80:20, A:B)
flow rate 1 mL/min
sample 1 mg/mL in methanol
injection 10 µL
detector UV, 205 nm
Description
Categories Analytical Chromatography, Chiral Applications, Anesthetics, Pharmaceutical (small molecule), All Pharmaceutical Applications, Anaesthetics