HPLC Analysis of Caffeine and Taurine on Ascentis® Express HILIC Column using Evaporative Light Scattering Detection (ELSD, Fast Analysis)
HPLC Analysis of Caffeine and Taurine on Ascentis® Express HILIC Column using Evaporative Light Scattering Detection (ELSD, Fast Analysis)
Conditions
column Ascentis Express HILIC, 5 cm x 3.0 mm I.D., 2.7 µm particles (53967-U)
column temp. 25 °C
mobile phase [A] 100 mM ammoniumformate, pH 3.0 with formic acid; [B] acetonitrile; (15:85, A:B)
flow rate 0.6 mL/min
sample 1000 µg/mL in 10:90, water: acetonitrile
injection 2 µL
detector ELSD, 55 °C, 3.5 bar nitrogen
Description
Analysis Note This application demonstrates the suitability of Ascentis Express HILIC for the efficient separation of caffeine and taurine. Elution is by aqueous normal phase (ANP) as an increase in organic will increase retention.
Categories Analytical Chromatography, StiµLants, Organic Acids, Drugs of Abuse
Featured Industry Vitamins, Nutraceuticals, and Natural Products