HPLC Analysis of Clenbuterol Enantiomers on Astec® CHIROBIOTIC® T2
HPLC Analysis of Clenbuterol Enantiomers on Astec® CHIROBIOTIC® T2
Conditions
column CHIROBIOTIC T2, 25 cm x 4.6 mm I.D., 5 µm particles (16024AST)
column temp. 25 °C
mobile phase 0.1 wt% ammonium trifluoroacetic acid in methanol
flow rate 1 mL/min
sample 1 mg/mL in methanol
injection 5 µL
detector UV, 230 nm
Description
Categories Analytical Chromatography, Forensics and Toxicology, Chiral Applications, bronchodilators, beta-agonists, drugs of abuse, Bronchodilators, Pharmaceutical (small molecule), Clinical, Forensic & Toxicology, Drugs of Abuse, All Pharmaceutical Applications
Featured Industry Forensics and Toxicology
Pharmaceutical (small molecule)