HPLC Analysis of Codeine and Metabolite on Ascentis® C18
HPLC Analysis of Codeine and Metabolite on Ascentis® C18
Conditions
column Ascentis C18, 15 cm x 4.6 mm I.D., 5 µm particles (581324-U)
column temp. 35 °C
mobile phase [A] 10 mM monobasic potassium phosphate (pH 3.0 with phosphoric acid); [B] acetonitrile; (90:10, A:B)
flow rate 1 mL/min
sample 50 µg/mL each in mobile phase
injection 10 µL
detector UV, 220 nm
Description
Categories Analytical Chromatography, Clinical, Forensics and Toxicology, Pharmaceutical Analysis, analgesics, Analgesics
Featured Industry Clinical
Forensics and Toxicology