HPLC Analysis of Coumachlor Enantiomers on Astec® CYCLOBOND® I 2000 DM
HPLC Analysis of Coumachlor Enantiomers on Astec® CYCLOBOND® I 2000 DM
Conditions
column CYCLOBOND I 2000 DM, 25 cm x 4.6 mm I.D., 5 µm particles (20924AST)
column temp. 25 °C
mobile phase [A] acetonitrile; [B] 20 mM ammonium acetate, pH 2.9; (20:80, A:B)
flow rate 0.8 mL/min
sample coumachlor, 1 mg/mL in acetonitrile:water (50:50)
injection 3 µL
detector UV, 230 nm
Description
Categories Analytical Chromatography, Pesticides & Herbicides
Featured Industry Chemicals and Industrial Polymers
Legal Information Astec is a registered trademark of Sigma-Aldrich Co. LLC, CYCLOBOND is a registered trademark of Sigma-Aldrich Co. LLC