HPLC Analysis of Creatine Kinase on Discovery® BIO PolyMA-WAX
HPLC Analysis of Creatine Kinase on Discovery® BIO PolyMA-WAX
Conditions
column Discovery BIO PolyMA-SCX, 5 cm x 4.6 mm, 5 µm particles (59601-U)
column temp. 25 °C
mobile phase [A] 20 mM sodium phosphate, pH 7.0; [B] 20 mM sodium phosphate, 0.5M NaCl, pH 7.0
flow rate 0.5 mL/min
injection 10 µL
detector UV, 280 nm
Description
Analysis Note Discovery BIO PolyMA-SCX and PolyMA-WAX columns give sharp, efficient peaks for a wide variety of proteins.
Categories Analytical Chromatography, Proteins
Featured Industry Life Science and Biopharma
Legal Information Discovery is a registered trademark of Sigma-Aldrich Co. LLC