HPLC Analysis of Enilconazol (Imazalil) Enantiomers on Astec® CYCLOBOND® I 2000
HPLC Analysis of Enilconazol (Imazalil) Enantiomers on Astec® CYCLOBOND® I 2000
Conditions
column CYCLOBOND I 2000, 25 cm x 4.6 mm I.D., 5 µm particles (20024AST)
column temp. 23 °C
mobile phase [A] 0.1% triethylamine acetate, pH 4.1; [B] acetonitrile; (90:10, A:B)
flow rate 0.7 mL/min
sample 5 mg/mL in methanol
injection 1 µL
detector UV, 230 nm
Description
Categories Analytical Chromatography, Chiral Applications, Pesticides & Herbicides, Environmental, Pesticides, All Environmental Applications
Featured Industry Agriculture
Legal Information Astec is a registered trademark of Sigma-Aldrich Co. LLC, CYCLOBOND is a registered trademark of Sigma-Aldrich Co. LLC