HPLC Analysis of Enkephalin Derivative (DAMME) on Discovery® HS F5
HPLC Analysis of Enkephalin Derivative (DAMME) on Discovery® HS F5
Conditions
column Discovery HS F5, 10 cm x 2.1 mm I.D., 5 µm particles (568501-U)
column temp. 22 °C
mobile phase [A] 25 mM HCO2H in water; [B] 25 mM HCO3H in acetonitrile
flow rate 0.2 mL/min
sample 10 mg/L in 0.1% HCO2H
injection 2 µL
detector ESI (+), full scan
Description
Categories Analytical Chromatography, Clinical, peptides, Analgesics
Featured Industry Clinical
Life Science and Biopharma
Legal Information Discovery is a registered trademark of Sigma-Aldrich Co. LLC