HPLC Analysis of Fenofibric Acid Enantiomers on Astec® CYCLOBOND® I 2000
HPLC Analysis of Fenofibric Acid Enantiomers on Astec® CYCLOBOND® I 2000
Conditions
column CYCLOBOND I 2000, 25 cm x 4.6 mm I.D., 5 µm particles (20024AST)
column temp. 22 °C
mobile phase [A] 0.1% triethylamine acetate, pH 4.0; [B] acetonitrile; (20:80, A:B)
flow rate 1 mL/min
sample 3 mg/mL in methanol
injection 1 µL
detector UV, 254 nm
Description
Categories Analytical Chromatography, Chiral Applications, Pharmaceutical (small molecule), Anticholesterolemics, Anticholesterol Drugs, All Pharmaceutical Applications
Featured Industry Pharmaceutical (small molecule)
Legal Information Astec is a registered trademark of Sigma-Aldrich Co. LLC, CYCLOBOND is a registered trademark of Sigma-Aldrich Co. LLC