HPLC Analysis of Fenoprofen Methyl Ester Enantiomers on Astec® CHIROBIOTIC® V
HPLC Analysis of Fenoprofen Methyl Ester Enantiomers on Astec® CHIROBIOTIC® V
Conditions
column CHIROBIOTIC V, 25 cm x 4.6 mm I.D., 5 µm particles (11024AST)
column temp. 23 °C
mobile phase [A] 20 mM sodium citrate, pH 6.3; [B] THF; (90:10, A:B)
flow rate 1 mL/min
sample 5 mg/mL in methanol
injection 5 µL
detector UV, 254 nm
Description
Categories Analytical Chromatography, Chiral Applications, Anti-inflammatories, Pharmaceutical (small molecule), All Pharmaceutical Applications