HPLC Analysis of Ferredoxin NADP Reductase on TSKgel® Phenyl-5PW
HPLC Analysis of Ferredoxin NADP Reductase on TSKgel® Phenyl-5PW
Conditions
column TSKgel Phenyl-5PW column, 7.5 cm x 7.5 mm I.D., 10 µm particles (807573)
column temp. 25 °C
mobile phase 60 min decreasing linear gradient from 1.5M to 0M ammonium sulfate in 0.1M sodium phosphate buffer, pH7
flow rate 0.5 mL/min
injection 3mg (crude) or 0.2mg (standard)
detector UV, 280 nm
Description
Categories Analytical Chromatography, Proteins
Featured Industry Life Science and Biopharma
Legal Information TSKgel is a registered trademark of Tosoh Corporation